HOBO® 2× External Temperature Data Logger
Supplier: ONSET COMPUTER CORPORATION SE
U23-003
470323-514EA 287.89 USD
470323-514
HOBO® 2× External Temperature Data Logger
Data Loggers
A weatherproof data logger with two external soil/water temperature probes on 6' cables for fast sensor response and deployment in tight spaces.
  • Weatherproof housing for use in outdoor or condensing environments
  • High accuracy
  • Small-diameter external sensor versions for measurements in tight spaces
  • Optic USB interface for fast and reliable download
Compatible with HOBOware® and HOBOware® Pro software for logger setup, graphing, and analysis.
Ordering information: Requires HOBOware® software and a USB cable. (NOTE: A USB cable is included with HOBOware® Pro CD or USB software purchase). HOBOware® Pro is required when using the U-DTW-1 waterproof shuttle.