Mounting Media Acrymount™ Plus, Statlab
Supplier: StatLab
SL80PLUS-16
SL80PLUS-4
76548-256EA
50.17
USD
76548-256
76548-258
Mounting Media Acrymount™ Plus, Statlab
Stains
Microscopy and staining reagents
A rapid-drying, toluene-based mounting medium/liquid coverglass. Compatible with xylene, and xylene-substitute clearing agents including d-Limonene.
- Slides mounted with Acrymount™ Plus may be screened in as little as 10 minutes.
- Formulated to reduce oxidation, preventing yellowing and fading in H&E, IHC and special stains after coverslipping
- Maintains an optimal refractive index for viewing and photo microscopy
- For labs seeking to eliminate final clearing with xylene, tissue may be coverslipped directly from absolute Isopropyl Alcohol
- Compatible with both manual and most automated coverslipping processes.
Part of the StatLab family of liquid mounting media.
Manufactured in the USA.