8-Bromoisoquinoline 97%

Supplier: Thermo Scientific Chemicals
Danger

45876-0010 45876-0050
TS45876-0010EA 68 USD
TS45876-0010 TS45876-0050
8-Bromoisoquinoline 97%
8-Bromoisoquinoline
Formula: C₉H₆BrN
MW: 208.0600 g/mol
Melting Pt: 78…81 °C
Storage Temperature: Ambient
MDL Number: MFCD04973298
CAS Number: 63927-22-0
UN: 2811
ADR: 6.1 III

Order Now

Specification Test Results

Appearance White to light brown Powder or crystalline powder
Infrared spectrum Conforms
Melting point 74 to 82 °C
GC ≥96.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR