2-Amino-4-chlorobenzonitrile 99%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 5-Chloro-2-cyanoaniline

45075-0100 45075-0500
TS45075-0100EA 60 USD
TS45075-0100 TS45075-0500
2-Amino-4-chlorobenzonitrile 99%
2-Amino-4-chlorobenzonitrile
Formula: C₇H₅ClN₂
MW: 152.58
Melting Pt: 159…162 °C
Storage Temperature: Ambient
MDL Number: MFCD00035927
CAS Number: 38487-86-4
UN: 3439
ADR: 6.1 III

Order Now

Specification Test Results

Appearance Off-white to yellow Powder
Infrared spectrum Conforms
Melting point 157 to 162 °C
Water ≤0.5 %
HPLC ≥98.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR