1-Iodo-4-(trifluoromethoxy)benzene 97%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
4-(Trifluoromethoxy)iodobenzene
44341-0010
44341-0050
TS44341-0010EA
35
USD
TS44341-0010
TS44341-0050
1-Iodo-4-(trifluoromethoxy)benzene 97%
1-Iodo-4-(trifluoromethoxy)benzene
Formula:
C₇H₄F₃IO Boiling Pt: 72…74 °C (15 mmHg) Density: 1.84 g/cm³ Flash Pt: 78 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD00042411 CAS Number: 103962-05-6 |
Specification Test Results
Appearance | Colorless to light yellow Liquid |
Infrared spectrum | Conforms |
GC | ≥96.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...