2,4,6-Trifluorophenylboronic acid 95%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: (2,4,6-Trifluorophenyl)metaboric acid, 2,4,6-Trifluorobenzeneboronic acid, Dihydroxy(2,4,6-trifluorophenyl)borane

43359-0010 43359-0050
TS43359-0010EA 41 USD
TS43359-0010 TS43359-0050
2,4,6-Trifluorophenylboronic acid 95%
2,4,6-Trifluorophenylboronic acid
Formula: C₆H₄BF₃O₂
Melting Pt: 239.5…247.5 °C
Storage Temperature: Ambient
MDL Number: MFCD01863169
CAS Number: 182482-25-3

Order Now

Specification Test Results

Appearance White to off-white Crystalline powder
Infrared spectrum Conforms
HPLC ≥94.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR