3-Chlorophenyl acetylene 97%

Supplier: Thermo Scientific Chemicals
Warning

43313-0010 43313-0050
TS43313-0010EA 58 USD
TS43313-0010 TS43313-0050
3-Chlorophenyl acetylene 97%
3-Chlorophenyl acetylene
Formula: C₈H₅Cl
Boiling Pt: 178…179 °C (760 mmHg)
Flash Pt: 63 °C
Storage Temperature: Refrigerator
MDL Number: MFCD03839984
CAS Number: 766-83-6

Order Now

Specification Test Results

Appearance Clear colorless to yellow Liquid
Infrared spectrum Conforms
GC ≥96.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR