4-Bromoaniline 96% pract.

Supplier: Thermo Scientific Chemicals
Danger

40287-1000 40287-5000
TS40287-1000EA 82 USD
TS40287-1000 TS40287-5000
4-Bromoaniline 96% pract.
4-Bromoaniline
Formula: C₆H₆BrN
MW: 172.02 g/mol
Boiling Pt: 223
Melting Pt: 60…64 °C
Density: 1.497 g/cm³
Flash Pt: >110 °C
Storage Temperature: Ambient
MDL Number: MFCD00007822
CAS Number: 106-40-1
EINECS: 203-393-9
UN: 2811
ADR: 6.1 III
Merck Index: 13,01388

Order Now

Specification Test Results

Appearance Cream to grey to brown Crystalline powder or crystals and/or chunks
Infrared spectrum Conforms
Melting point 60 to 65 °C
GC ≥95 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR