3,3',4,4'-Benzophenonetetracarboxylic dianhydride 97+%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
Benzophenone-3,3',4,4'-tetracarboxylic dianhydride
40193-0025
40193-5000
TS40193-0025EA
363
USD
TS40193-0025
TS40193-5000
3,3',4,4'-Benzophenonetetracarboxylic dianhydride 97+%
3,3',4,4'-Benzophenonetetracarboxylic dianhydride
Formula:
C₁₇H₆O₇ MW: 322.23 g/mol Melting Pt: 219…226 °C Storage Temperature: Ambient |
MDL Number:
MFCD00005923 CAS Number: 2421-28-5 EINECS: 219-348-1 |
Specification Test Results
Appearance | Yellow to tan Powder |
Infrared spectrum | Conforms |
Melting point | 220 to 229 °C |
Free acid | ≤2.2 % |
Titration by coupling | ≥97.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...