Anthrone 95%, pure
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
Anthracen-9(10H)-one
40136-0250
40136-1000
40136-5000
TS40136-0250EA
90
USD
TS40136-0250
TS40136-1000
TS40136-5000
Anthrone 95%, pure
Anthrone
Formula:
C₁₄H₁₀O MW: 194.23 g/mol Boiling Pt: 721 °C (760 mmHg) Melting Pt: 152…157 °C Storage Temperature: Ambient |
MDL Number:
MFCD00001187 CAS Number: 90-44-8 |
Specification Test Results
Appearance | Yellow to beige Crystalline powder or needles |
Infrared spectrum | Conforms |
Melting point | 155 to 161 °C |
GC | ≥94 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...