2-Amino-5-nitrophenol 95%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 2-Hydroxy-4-nitroaniline

40090-0010 40090-0250 40090-0500 40090-2500
TS40090-0010EA 483 USD
TS40090-0010 TS40090-0250 TS40090-0500 TS40090-2500
2-Amino-5-nitrophenol 95%
2-Amino-5-nitrophenol
Formula: C₆H₆N₂O₃
Melting Pt: 198…202 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00007692
CAS Number: 121-88-0
EINECS: 204-503-8

Order Now

Specification Test Results

Appearance Rust brown to brown Crystalline powder
Infrared spectrum Conforms
Titration with HClO4 ≥94 %
GC ≥96.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR