1-Fluoro-4-ethynylbenzene 98%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 1-Ethynyl-4-fluorobenzene, (4-Fluorophenyl)acetylene, (4-Fluorophenyl)ethyne

37137-0010 37137-0050
TS37137-0010EA 51 USD
TS37137-0010 TS37137-0050
1-Fluoro-4-ethynylbenzene 98%
1-Fluoro-4-ethynylbenzene
Formula: C₈H₅F
MW: 120.13
Boiling Pt: 55…56 °C (40 mmHg)
Melting Pt: 27…28 °C
Density: 1.048 g/cm³
Flash Pt: 29 °C
Storage Temperature: Ambient
MDL Number: MFCD00168823
CAS Number: 766-98-3
UN: 1325
ADR: 4.1 III

Order Now

Specification Test Results

Appearance Clear colorless to yellow Liquid after melting
Infrared spectrum Conforms
Refractive index 1.5155 to 1.5185 (20°C, 589 nm)
GC ≥97.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR