α-Chlorothioanisole 97%

Supplier: Thermo Scientific Chemicals

Synonyms: Chloromethyl phenyl sulphideChloromethyl phenyl sulfide

20236-0250 20236-0050
TS20236-0050EA 71 USD
TS20236-0050 TS20236-0250
α-Chlorothioanisole 97%
α-Chlorothioanisole
Formula: C₇H₇ClS
Boiling Pt: 66 °C (.3hPa)
Density: 1.23 g/cm³
Flash Pt: 101 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00000921
CAS Number: 7205-91-6

Order Now

Specification Test Results

Appearance Clear colorless to light yellow Liquid
Infrared spectrum Conforms
GC ≥96.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR