Buffers, running buffers for Biacore Systems, Cytiva
Supplier: Cytiva
Buffers developed and verified to work in Biacore™ systems.
The recommended running buffer for an assay depends on the type of molecules used in the interaction, which assay will be run, and the type of sensor chip used. Supplied in ready to use or concentrated form.