NEBNext FFPE DNA Repair v2 Module, New England Biolabs
Supplier: New England Biolabs (NEB)
E7360L
E7360S
76559-400EA
641
USD
76559-400
76559-398
NEBNext FFPE DNA Repair v2 Module, New England Biolabs
Nucleic Acid Reagents
Next Generation Sequencing Reagents
This new product further improves repair of FFPE DNA in next generation sequencing (NGS) workflows. The methods used for fixation and storage of Formalin-Fixed, Paraffin-Embedded (FFPE) DNA samples cause significant damage, making it challenging to obtain high quality sequence data.
- Higher repair efficiency with FFPE DNA
- A more streamlined workflow for NGS library preparation
- Convenient reaction buffer for efficient FFPE DNA repair, end repair and dA-tailing
The NEBNext FFPE DNA Repair v2 Module is an optimized cocktail of enzymes designed to repair FFPE DNA, and supplied with optimized reagents to enable a streamlined workflow for NGS library preparation.