Labclean™ Cleanroom Wipes, 100% Polyester, United Scientific Supplies
Supplier: United Scientific Supplies
PWIPE12X12 PWIPE4X4 PWIPE9X9
470340-094EA 71.3 USD
470340-094 470339-504 470339-168
Labclean™ Cleanroom Wipes, 100% Polyester, United Scientific Supplies
Wipes Cleanroom Wipes Dry Cleanroom Wipes
Polyester wipes for cleanroom environments.
  • 100% virgin, continuous filament, double-knit polyester fiber
  • Laser cut sealed edge for reduced fiber contamination
  • Moderate absorbency and abrasion resistance
  • Low levels of particulate and extractable counts
Polyester wipes are 100% virgin, continuous-filament polyester in a double-knit, no-run interlock design with a laser cut sealed edge. Recommended for use in cleanroom critical environments and a wide variety of applications including cleaning medical devices, electronics displays, instrument panels, semiconductors, and microelectronics products. Laundered and packaged in a cleanroom. Vacuum-packed in solvent-safe, cleanroom-compatible polyethylene with an inner bag label. Available in three convenient sizes.