Pipette Filter Tips, Low Retention, Racked, Sterile, United Scientific Supplies
Supplier: United Scientific Supplies
P11065 P11073 P11077
470339-552EA 140.6 USD
470339-552 470339-702 470339-056
Pipette Filter Tips, Low Retention, Racked, Sterile, United Scientific Supplies
Pipette Tips
Sterile and disposable low retention pipette filter tips.
  • Graduated
  • Free of DNase, RNase and pyrogen
  • Tips are sterile and autoclavable
  • Non-cytotoxic, non-hemolytic, and have a 99.9% bacterial filtration efficiency (BFE)
Low Retention Micropipette Tips racked are disposable tips intended for use with most brands of pipettors. Manufactured from 100% pure porous polyethylene, filters are designed to prevent liquids and aerosols from entering the body of the micropipette to protect against cross-contamination.
Best suited for biological applications, these tips are. Deliver 10X lower retention levels with viscous samples, as compared to standard pipette tips, Tips are manufactured from ultra-pure USP Class VI certified, virgin polypropylene.