Labclean™ Cleanroom Wipes, 55% Cellulose/45% Polyester, United Scientific Supplies
Supplier: United Scientific Supplies
CPWIPE12X12 CPWIPE4X4 CPWIPE9X9
470339-888EA 91.8 USD
470339-888 470340-002 470340-100
Labclean™ Cleanroom Wipes, 55% Cellulose/45% Polyester, United Scientific Supplies
Wipes Cleanroom Wipes Dry Cleanroom Wipes
100% polyester wipes for use in cleanroom environments.
  • 100% virgin, continuous filament, double-knit polyester fiber
  • Laser cut, sealed edge for reduced fiber contamination
  • Moderate absorbency and abrasion resistance
  • Low levels of particulate and extractable counts
Spunlace non-woven wipes are constructed from a hydro-entangled, non-woven blend of 55% cellulose and 45% polyester. This composite substrate has the highly absorbent properties of a natural fiber with the cleanliness and strength of a synthetic yarn. Recommended for use in cleanroom Class 1More Product Information