(1S,4S)-2-Boc-2,5-diazabicyclo[2.2.1]heptane 95%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: tert-butyl (1S,4S)-2,5-diazabicyclo[2.2.1]heptane-2-carboxylate

39804-0050 39804-0010
TS39804-0050EA 676 USD
TS39804-0050 TS39804-0010
(1S,4S)-2-Boc-2,5-diazabicyclo[2.2.1]heptane 95%
(1S,4S)-2-Boc-2,5-diazabicyclo[2.2.1]heptane
Formula: C₁₀H₁₈N₂O₂
MW: 198.26 g/mol
Melting Pt: 74…76 °C
Storage Temperature: Ambient
MDL Number: MFCD01569250
CAS Number: 113451-59-5

Order Now

Specification Test Results

Appearance White to light beige Powder
Infrared spectrum Conforms
Melting point 73 to 77 °C
GC ≥94 %
Specific optical rotation -41° to -47° (20 °C, 589 nm) (C=1, chloroform)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR