2,6-Dichloronicotinic acid 98+%

Supplier: Thermo Scientific Chemicals
Warning

39661-0010 39661-0050 39661-0250
TS39661-0010EA 35 USD
TS39661-0010 TS39661-0050 TS39661-0250
2,6-Dichloronicotinic acid 98+%
2,6-Dichloronicotinic acid
Formula: C₆H₃Cl₂NO₂
Melting Pt: 147…150 °C
Storage Temperature: Ambient
MDL Number: MFCD00075583
CAS Number: 38496-18-3

Order Now

Specification Test Results

Appearance Off-white to light yellow Crystalline powder
Infrared spectrum Conforms
HPLC ≥98.0 %
Titration with NaOH ≥98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR