4-Aminophenylboronic acid pinacol ester 97%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 4-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)aniline, 4-Aminobenzeneboronic acid pinacol ester

36638-0010 36638-0050
TS36638-0010EA 67 USD
TS36638-0010 TS36638-0050
4-Aminophenylboronic acid pinacol ester 97%
4-Aminophenylboronic acid pinacol ester
Formula: C₁₂H₁₈BNO₂
Melting Pt: 165…170 °C
Storage Temperature: Refrigerator
MDL Number: MFCD02093721
CAS Number: 214360-73-3

Order Now

Specification Test Results

Appearance White to light beige Crystalline powder
Infrared spectrum Conforms
Melting point 165 to 170 °C
GC ≥96.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR