2,5-Dibromo-4-picoline 98%

Supplier: Thermo Scientific Chemicals
Warning

36105-0010 36105-0050
TS36105-0010EA 35 USD
TS36105-0010 TS36105-0050
2,5-Dibromo-4-picoline 98%
2,5-Dibromo-4-picoline
Formula: C₆H₅Br₂N
Melting Pt: 37…41 °C
Storage Temperature: Ambient
MDL Number: MFCD00234955
CAS Number: 3430-26-0
UN: 2811
ADR: 6.1,III

Order Now

Specification Test Results

Appearance Light yellow to orange Low melting solid
Infrared spectrum Conforms
Melting point 37 to 41 °C
GC ≥97.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR