3-Amino-2-pyridone 97%
Supplier: Thermo Scientific Chemicals
|
Warning
|
36071-0010
36071-0050
TS36071-0010EA
36
USD
TS36071-0010
TS36071-0050
3-Amino-2-pyridone 97%
3-Amino-2-pyridone
Formula:
C₅H₆N₂O Melting Pt: 118…130 °C Storage Temperature: Ambient |
MDL Number:
MFCD03411556 CAS Number: 33630-99-8 |
Specification Test Results
Appearance | Dark brown to black Powder |
Infrared spectrum | Conforms |
HPLC | ≥96.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...