Succinimide 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
Pyrrolidine-2,5-dione
34865-0050
34865-1000
34865-5000
TS34865-0050EA
38
USD
TS34865-0050
TS34865-1000
TS34865-5000
Succinimide 98%
Succinimide
Formula:
C₄H₅NO₂ Boiling Pt: 285…290 °C (760 mmHg) Melting Pt: 121…126 °C Flash Pt: 201 °C Storage Temperature: Ambient |
MDL Number:
MFCD00005495 CAS Number: 123-56-8 EINECS: 204-635-6 Merck Index: 12,09040 |
Specification Test Results
Appearance | Off-white to beige to light brown Powder or flakes |
Infrared spectrum | Conforms |
Melting point | 121 to 126 °C |
Titration with NaOH | ≥97.5 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...