2-Bromo-5-methylpyridine 98%

Supplier: Thermo Scientific Chemicals
Warning

34717-0050 34717-0250
TS34717-0050EA 57 USD
TS34717-0050 TS34717-0250
2-Bromo-5-methylpyridine 98%
2-Bromo-5-methylpyridine
Formula: C₆H₆BrN
Boiling Pt: 95…96 °C (12.5 mmHg)
Melting Pt: 40…45 °C
Flash Pt: 103 °C
MDL Number: MFCD00209553
CAS Number: 3510-66-5

Order Now

Specification Test Results

Appearance White to light yellow to light brown Low melting crystalline solid or crystals
Infrared spectrum Conforms
Melting point 40 to 45 °C
GC ≥97.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR