Azetidine 98%
Supplier: Thermo Scientific Chemicals
|
Danger
|
29673-0050
29673-2500
29673-0010
TS29673-2500EA
139
USD
TS29673-2500
TS29673-0010
TS29673-0050
Azetidine 98%
Azetidine
Formula:
C₃H₇N Boiling Pt: 61…62 °C (760 mmHg) Density: 0.85 g/cm³ Flash Pt: –20 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD00005165 EINECS: 207-963-8 UN: 2733 ADR: 3,II |
Specification Test Results
Appearance | Clear colorless Liquid |
Infrared spectrum | Conforms |
Refractive index | 1.4290 to 1.4330 (20°C, 589 nm) |
GC | ≥97.5 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...