Tetrathiafulvalene 99+%

Supplier: Thermo Scientific Chemicals

17941-0010 17941-2500
TS17941-0010EA 190 USD
TS17941-0010 TS17941-2500
Tetrathiafulvalene 99+%
Tetrathiafulvalene
Formula: C₆H₄S₄
Melting Pt: 117…120 °C
Storage Temperature: Ambient
MDL Number: MFCD00005492
CAS Number: 31366-25-3
EINECS: 250-593-7

Order Now

Specification Test Results

Appearance Orange to brown Crystalline powder or crystals
Infrared spectrum Conforms
Melting point 117 to 120 °C
GC ≥99.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR