2-Amino-3,5-dichloropyridine 97%

Supplier: Thermo Scientific Chemicals
Warning

15938-0050 15938-0250 15938-1000
TS15938-0050EA 14 USD
TS15938-0050 TS15938-0250 TS15938-1000
2-Amino-3,5-dichloropyridine 97%
2-Amino-3,5-dichloropyridine
Formula: C₅H₄Cl₂N₂
Melting Pt: 80…82 °C
MDL Number: MFCD00006313
CAS Number: 4214-74-8
EINECS: 224-143-5

Order Now

Specification Test Results

Appearance Light beige to light brown Powder
Infrared spectrum Conforms
Melting point 79 to 83 °C
Water ≤0.5 %
HPLC ≥96.0 %
Acid insoluble substances ≤0.05 % (in HCl)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR