1-Methylpyrrolidine 98%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: N-Methylpyrrolidine

12761-1000 12761-0025 12761-5000
TS12761-0025EA 615 USD
TS12761-0025 TS12761-1000 TS12761-5000
1-Methylpyrrolidine 98%
1-Methylpyrrolidine
Formula: C₅H₁₁N
MW: 85.15 g/mol
Boiling Pt: 80…81 °C (760 mmHg)
Melting Pt: –90 °C
Density: 0.819 g/cm³
Flash Pt: –21 °C
Storage Temperature: Ambient
MDL Number: MFCD00003173
CAS Number: 120-94-5
EINECS: 204-438-5
UN: 3286
ADR: 3,II

Order Now

Specification Test Results

Appearance Clear colorless Liquid
Infrared spectrum Conforms
Refractive index 1.4230 to 1.4260 (20°C, 589 nm)
GC ≥97.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR