Furfuryl alcohol 98%
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
Furfurol
11979-0010
11979-0100
11979-0025
11979-2500
TS11979-0025EA
112
USD
TS11979-0025
TS11979-2500
TS11979-0100
TS11979-0010
Furfuryl alcohol 98%
Furfuryl alcohol
Formula:
C₅H₆O₂ MW: 98.1 g/mol Boiling Pt: 170 °C (760 mmHg) Melting Pt: –29 °C Density: 1.13 g/cm³ Flash Pt: 65 °C Storage Temperature: Ambient |
MDL Number:
MFCD00003252 CAS Number: 98-00-0 EINECS: 202-626-1 UN: 2874 ADR: 6.1,III |
Specification Test Results
Appearance | Clear yellow Liquid (May darken on storage) |
Infrared spectrum | Conforms |
Refractive index | 1.4850 to 1.4880 (20°C, 589 nm) |
Water | ≤0.3 % |
GC | ≥97.5 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...