Dibenzothiophene 98%

Supplier: Thermo Scientific Chemicals
Warning

11254-0010 11254-0250 11254-1000 11254-2500
TS11254-0010EA 919 USD
TS11254-0010 TS11254-0250 TS11254-1000 TS11254-2500
Dibenzothiophene 98%
Dibenzothiophene
Formula: C₁₂H₈S
Boiling Pt: 332…333 °C (760 mmHg)
Melting Pt: 97…100 °C
Flash Pt: 170 °C
Storage Temperature: Ambient
MDL Number: MFCD00004969
CAS Number: 132-65-0
EINECS: 205-072-9
UN: 3077
ADR: 9,III

Order Now

Specification Test Results

Appearance White to beige to grey Powder or crystals and/or chunks
Infrared spectrum Conforms
Melting point 97 to 101 °C
GC ≥96.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR