5-Amino-2-methoxypyridine 90% tech.
Supplier: Thermo Scientific Chemicals
|
Warning
|
10395-0050
10395-0250
10395-1000
TS10395-0050EA
53
USD
TS10395-0050
TS10395-0250
TS10395-1000
5-Amino-2-methoxypyridine 90% tech.
5-Amino-2-methoxypyridine
Formula:
C₆H₈N₂O MW: 124.14 Boiling Pt: 265 Melting Pt: 29…31 °C Density: 1.575 g/cm³ Flash Pt: >110 °C Storage Temperature: Ambient |
MDL Number:
MFCD00006264 CAS Number: 6628-77-9 EINECS: 229-612-8 |
Specification Test Results
Appearance | Red to yellow-brown Liquid |
Infrared spectrum | Conforms |
GC | ≥88 % |
Clarity of solution | Passes test |
Specific optical rotation | +103° to +108° (20°C, 589 nm) (c=5, H2O) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...