Mortar and Pestle Set, Porcelain, United Scientific Supplies
Supplier: United Scientific Supplies
JMD275 JMD050 JMD400 JMD150 JMD070 JMD1900 JMD750
470339-254EA 29.8 USD
470339-254 470339-596 470339-372 470339-296 470340-276 470339-846 470340-046
Mortar and Pestle Set, Porcelain, United Scientific Supplies
Mortars and Pestles Mortars with Pestles
Deep form mortar with oversized pestle.
  • Mortar features pour spout and is glazed outside (excluding bottom), unglazed inside
  • Oversized pestle is glazed to grinding surface
Deep-form mortar with pour spout. Glazed on the outside (except for the bottom) and unglazed inside. Oversized pestle is glazed to grinding surface.