Measuring Cylinder, PMP, Class A, United Scientific Supplies
Supplier: United Scientific Supplies
P50507 P50508 P50501 P50502 P50503 P50504 P50505 P50506
470339-628EA 112.5 USD
470339-628 470340-180 470339-334 470340-232 470339-138 470339-358 470340-092 470340-166
Measuring Cylinder, PMP, Class A, United Scientific Supplies
Cylinders Measuring Cylinders
Clear graduated cylinders with large spouts for smooth pouring.
  • Polymethylpentene cylinder
  • Glass-like clarity, with easy-to-read molded in graduations
  • Meets accuracy requirements of Class A under DIN 12681; conformity test certificate indicates batch number and year of production
  • Autoclavable
Polymethylpentene cylinder with glass-like clarity, with easy-to-read molded in graduations.
Features hexagon-shaped base for extra stability and large spout for smooth flow of liquids during pouring. Autoclavable to 121 °C.
Meets accuracy requirements of Class A under DIN 12681. Supplied with conformity test certificate that indicates batch number and year of production.