Jade Bottle Top Dispenser, United Scientific Supplies
Supplier: United Scientific Supplies
BTJD10 BTJD100 BTJD2.5 BTJD30 BTJD5 BTJD60
470339-972EA 545.6 USD
470339-972 470340-144 470339-442 470339-930 470339-774 470339-670
Jade Bottle Top Dispenser, United Scientific Supplies
Bottle Top Dispensers
Precise dispensing for a wide range of reagents.
  • Fully autoclavable bottle top dispenser
  • Includes adapters to fit a wide range of bottle sizes: 28, 32, 38, 40 and 45 mm
  • Features re-circulation valve to redirect liquid back into the bottle without reagent loss
  • Smooth plunger movement enables bubble-free dispensing
Five adapters are included with each unit for use with a wide range of bottle sizes. Adapters included are: 28, 32, 38 , 40 and 45 mm.
Fully autoclavable dispenser features a re-circulation valve that redirects liquid back into the bottle without the loss of reagent. Operates with a smooth plunger movement for bubble-free dispensing and effortless performance, even under demanding laboratory conditions.