Bottle Top Dispenser, United Scientific Supplies
Supplier: United Scientific Supplies
BTDR-6 BTDR-1 BTDR-2 BTDR-3 BTDR-4 BTDR-5
470340-084EA 508.4 USD
470340-084 470339-216 470339-180 470339-086 470339-962 470339-642
Bottle Top Dispenser, United Scientific Supplies
Bottle Top Dispensers
Precise dispensing for laboratory solutions.
  • Includes adapters to fit a wide range of bottle sizes: 28, 30, 32, 36, 40 and 45 mm
  • All wetted parts made from PTFE and borosilicate glass for maximum chemical resistance
  • Not suitable for use with hydrogen fluoride
  • Individually calibrated and certified in an ISO 17025 accredited laboratory in accordance with ISO 8655 standards
  • Calibration certificate included
Five adapters are included with each unit for use with a wide range of bottle sizes. All wetted parts are made from PTFE and borosilicate glass forMore Product Information