Black Diamond Analog Heating Mantles, United Scientific Supplies
Supplier: United Scientific Supplies
UNANHMTL-500 UNANHMTL-5000 UNANHMTL-100 UNANHMTL-1000 UNANHMTL-2000 UNANHMTL-250 UNANHMTL-3000
470339-870EA 421.3 USD
470339-870 470339-308 470339-038 470340-224 470340-008 470339-348 470339-058
Black Diamond Analog Heating Mantles, United Scientific Supplies
Heaters Heating Mantles
Durable heating system for flasks of all sizes.
  • Stainless steel and aluminum construction
  • Glass fiber yarn knitted mantle provides uniform heating to round bottom flasks
  • Unique spill collect system
  • Easy to use and maintain
  • Max. temperature: 400 °C
This series of analog heating mantles includes a case constructed of durable stainless steel and aluminum materials. The temperature level is adjustable up to 400 °C. Knitted mantle is made from glass fiber yarn and designed to securely hold round bottom flasks with the top half of the flask visible during operation. Available in various sizes to hold flasks from 100 to 5000 ml capacity. Includes a spill collect system to drain solution if a flask breaks during use.