High Alumina Crucibles, Conical Form, United Scientific Supplies
Supplier: United Scientific Supplies
JAC020 JAC030 JAC050 JAC100
470339-960EA 41.5 USD
470339-960 470339-270 470340-234 470339-882
High Alumina Crucibles, Conical Form, United Scientific Supplies
Crucibles
Unglazed high alumiona crucibles.
  • Conical form crucible
  • Made from 99.7% pure aluminum oxide for maximum use temperature of 1750 °C (3182 °F), Temperature change rate should not exceed
  • 150 °C (302 °F) per hour
  • Recommended for use with refractory metals, suitable for glass melting
This conical-form crucible with 20 ml capacity is crafted from 99.7% pure aluminum oxide, ideal for high-temperature work demanding contamination-free results. Maximum use temperature is 1750 °C (3182 °F), temperature change rate should not exceed 150 °C (302 °F) per hour.
High-alumina products are highly refractory, meant for use in reducing and oxidizing atmospheres. Material is inert in hydrogen and carbonaceous atmospheres and offers high resistance to alkalies and other fluxes. Recommended for use with refractory metals. Suitable for glass melting, including borosilicate glass.