Paraffin 360, ParaPro™

Supplier: StatLab

Synonyms: Wax ParaffinParaffin wax

PP360
76548-192CS 152.8 USD
76548-192
Paraffin 360, ParaPro™
Paraffin

A low-melting paraffin appropriate for infiltration and embedding.


  • Ideal for standard ribbons down to the thinnest 2-micron sections
  • Formulated with a proprietary blend of low-molecular-weight paraffin and polymers
  • Melting point 50°-54°C
  • 0.8 kg bag, 10/cs


Part of the StatLab line of ParaPro paraffin.

Manufactured in the USA

CAS Number: 8002-74-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR