Paraffin LMP, ParaPro™
Supplier: StatLab
Synonyms:
Wax Paraffin
, Paraffin wax
PPLMP
76548-196CS
149.14
USD
76548-196
Paraffin LMP, ParaPro™
Paraffin
A medium/hard paraffin. For infiltration and embedding.
- Consists of a specialized blend of low-molecular-weight paraffin and polymers to reduce melting point
- The lower melting point protects tissues from excessive temperature during processing
- Melting point 50°-54° C
- 1 kg bag, 8/cs
Part of the StatLab line of ParaPro paraffin.
Manufactured in the USA
CAS Number:
8002-74-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...