Paraffin Blue, ParaPro™
Supplier: StatLab
Synonyms:
Wax Paraffin
, Paraffin wax
PPBLU
76548-194CS
134.15
USD
76548-194
Paraffin Blue, ParaPro™
Paraffin
Universal infiltration and embedding paraffin composed of light blue pellets. A medium paraffin.
- The blue color makes it easier to visualize small biopsies after processing
- Highly refined with the addition of synthetic polymers, enabling rapid impregnation
- Optimized for dermatology and urology specimens
- Melting point 54°-57° C
- 1 kg bag, 8/cs
Part of the StatLab line of ParaPro paraffin.
Manufactured in the USA
CAS Number:
8002-74-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...