Acetone ACS
Supplier: StatLab
|
Danger
|
Synonyms:
2-Propanone, DMK, Propanone, Dimethyl ketone
00960-1
00960-16
76547-968EA
98.58
USD
76547-968
76547-970
Acetone ACS
Acetone
Acetone, ACS Grade
- Reliable stain reagent to produce the results you need
Part of the StatLab Stain Reagent line.
Manufactured in the USA
Formula:
CH₃COCH₃ MW: 58.08 g/mol |
MDL Number:
MFCD00008765 CAS Number: 67-64-1 UN: 1090 ADR: 3,II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...