Papanicolaou's solution EA 65 stain for cytology
Supplier: StatLab
|
Danger
|
Synonyms:
EA 36
SL302
76548-226EA
53.82
USD
76548-226
Papanicolaou's solution EA 65 stain for cytology
Papanicolaou's solution EA 65
Alcohol-based, for routine cytoplasmic staining.
Part of the StatLab family of Cytology stains.
Manufactured in the USA
UN:
1987 ADR: 3,II |
Learn more
![](/m-us.vwr.com/en_US/images/about_vwr_275x315.jpg)