Treosin™
Supplier: StatLab
Danger
SL93-1 SL93-16
76540-470EA 158.25 USD
76540-470 76548-272
Treosin™
Treosin™
Slightly acidified combination of Eosin-Y with Orange-G and Acid Fuchsin.
  • Suitable as replace-ment for routine eosin
  • Distinct shades of eosin easily obtained from bright red-orange (red blood cell’s and fibrin deposits) to bright red (collagen fibers and skeletal muscle) to red-pink (smooth muscle)
  • In many cases Treosin™ could eliminate the need for trichrome staining
  • Excellent for frozen sections
Part of the StatLab Classic stain line.
Manufactured in USA.
UN: 1987
ADR: 3 II