Eosin Y alcoholic solution, Classic

Supplier: StatLab
Warning

Synonyms: Acid Red 87, Tetrabromofluorescein, 2-(3,6-Dihydroxy-2,4,5,7-tetrabromoxanthen-9-yl)-benzoic acid, 2,4,5,7-Tetrabromofluorescein, Eosin yellowish, Solvent red 43, Bromofluorescein, Bromo acid J. TS, XL, or XX, CI 45380, Bronze Bromo ES, Spiro[isobenzofuran-1(3H),9'-[9H]xanthen]-3-one, 2',4',5',7'-tetrabromo-3',6'-dihydroxy-

SL98-1 SL98-16
76540-472EA 146.91 USD
76540-472 76548-290
Eosin Y alcoholic solution, Classic
Eosin Y

Ready-to-use counterstain for routine H & E procedures.


  • Produces a brilliant counterstain with excellent differentiation or cytoplasmic details and non-nuclear tissue components


Part of the StatLab Classic stain line.

Manufactured in USA.

Formula: C₂₀H₈Br₄O₅
MW: 647.89 g/mol
Melting Pt: 295
MDL Number: MFCD00036189
CAS Number: 15086-94-9
UN: 1987
ADR: 3 III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR