Hematoxylin Gill 2, Classic
Supplier: StatLab
|
Danger
|
Synonyms:
DcH-48, Natural Black I, Hydroxybrasilin
SL94-1
SL94-16
76548-274EA
217.66
USD
76548-274
76548-276
Hematoxylin Gill 2, Classic
Hematoxylin staining solutions
Medium dye concentration for histology or intense cytology staining.
- Produces a darker nuclei, or used when shorter staining times are needed
- Nuclear detail is very distinct and has great clarity
Part of the StatLab Classic stain line.
Manufactured in USA.
Formula:
C₁₆H₁₄O₆ MW: 302.28 g/mol |
MDL Number:
MFCD00078111 CAS Number: 517-28-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...