Hematoxylin Harris, Classic
Supplier: StatLab
|
Danger
|
Synonyms:
DcH-48, Natural Black I, Hydroxybrasilin
SL90-1
SL90-16
76548-260EA
217.65
USD
76548-260
76548-262
Hematoxylin Harris, Classic
Hematoxylin staining solutions
Modified traditional formulation.
- Contains no mercury
- Generally used as a regressive stain, but may also be used in progressive staining protocols
- Produces rapid and distinct nuclear detail
Part of the StatLab Classic stain line.
Manufactured in USA.
Formula:
C₁₆H₁₄O₆ MW: 302.28 g/mol |
MDL Number:
MFCD00078111 CAS Number: 517-28-2 UN: 1993 ADR: 3 III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...