Hematoxylin, Select

Supplier: StatLab
Danger

Synonyms: HydroxybrasilinDcH-48Natural Black I

SL401
76548-230EA 76.19 USD
76548-230
Hematoxylin, Select
Hematoxylin staining solutions

Proprietary progressive nuclear stain, considered a hybrid hematoxylin product.


  • Possesses the rapid staining qualities of Gill Hematoxylins along with the familiar brilliant nuclear chromatin delineation of a Harris
  • Does not require daily filtering
  • Has no affinity for background or mucin staining
  • May be used for routine H&E staining, IHC counterstaining, frozen section staining


Part of the StatLab Select stain line.

Manufactured in USA.

Formula: C₁₆H₁₄O₆
MW: 302.28 g/mol
MDL Number: MFCD00078111
CAS Number: 517-28-2
UN: 1789
ADR: 8,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR