Eosin Y, Vintage

Supplier: StatLab
Warning

Synonyms: Acid Red 872,4,5,7-TetrabromofluoresceinEosin yellowishSolvent red 43BromofluoresceinSpiro[isobenzofuran-1(3H),9'-[9H]xanthen]-3-one, 2',4',5',7'-tetrabromo-3',6'-dihydroxy-CI 45380Tetrabromofluorescein2-(3,6-Dihydroxy-2,4,5,7-tetrabromoxanthen-9-yl)-benzoic acidBronze Bromo ESBromo acid J. TS, XL, or XX

SL101
76548-208EA 54.34 USD
76548-208
Eosin Y, Vintage
Eosin Y

Proprietary, alcoholic and acidified formulation


  • Produces a brilliant cytoplasmic stain with three distinct color variations in a short amount of staining time
  • Yields a very familiar cytoplasmic staining result


Part of the StatLab Vintage stain line.

Manufactured in USA.

Formula: C₂₀H₈Br₄O₅
MW: 647.89 g/mol
MDL Number: MFCD00036189
CAS Number: 15086-94-9
UN: 1987
ADR: 3,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR