Hematoxylin, Reserve
Supplier: StatLab
|
Warning
|
Synonyms:
Hydroxybrasilin
, DcH-48
, Natural Black I
SL200
76548-218EA
75.05
USD
76548-218
Hematoxylin, Reserve
Hematoxylin staining solutions
A uniquely formulated hematoxylin stain that can be used in place of Gill or Harris hematoxylins.
- Rapidly provides unsurpassed nuclear chromatin delineation with no affinity for background or mucin staining
- Does not require daily filtering or form metallic sheens
- May be used for both histology and cytology applications
- Engineered to only stain the nuclear chromatin
- No mucin staining is seen.
Part of the StatLab Reserve stain line.
Manufactured in USA.
Formula:
C₁₆H₁₄O₆ MW: 302.28 g/mol |
MDL Number:
MFCD00078111 CAS Number: 517-28-2 UN: 1789 ADR: 8,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...